ENA423210072 Enamine

2-cyano-N-[(4-nitrophenyl)methyl]acetamide

  • MDL number MFCD12763463
  • Cas Number : 566926-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172302
Availability details for
Quantity
  • Availability
  • Add to Cart