ENA423210073 Enamine

3-(prop-2-yn-1-yl)oxolane-2,5-dione

  • MDL number MFCD18432058
  • Cas Number : 98550-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172303
Availability details for
Quantity
  • Availability
  • Add to Cart