ENA423210087 Enamine

1-methyl-3-(3-methylbutyl)-1H-pyrazol-5-amine

  • MDL number MFCD11209672
  • Cas Number : 1152707-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172427
Availability details for
Quantity
  • Availability
  • Add to Cart