ENA423210088 Enamine

methyl 1,5-dioxaspiro[2.5]octane-2-carboxylate

  • MDL number MFCD21321609
  • Cas Number : 1495959-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172428
Availability details for
Quantity
  • Availability
  • Add to Cart