ENA423210091 Enamine

3-(2-bromoethoxy)-1,1,2,2-tetrafluoropropane

  • MDL number MFCD11197517
  • Cas Number : 1038730-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171969
Availability details for
Quantity
  • Availability
  • Add to Cart