ENA423210092 Enamine

1-(1-tert-butyl-5-methyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD00112453
  • Cas Number : 1152927-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-172436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172436
Availability details for
Quantity
  • Availability
  • Add to Cart