ENA423210098 Enamine

4-phenyl-1,3-oxazol-2-amine

  • MDL number MFCD13189375
  • Cas Number : 33119-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172459
Availability details for
Quantity
  • Availability
  • Add to Cart