ENA423210102 Enamine

5-methoxy-1H-indol-4-amine

  • MDL number MFCD18808975
  • Cas Number : 431046-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172479
Availability details for
Quantity
  • Availability
  • Add to Cart