ENA423210112 Enamine

3H-imidazo[4,5-b]pyridin-2-ylmethanamine dihydrochloride

  • MDL number MFCD09038189
  • Cas Number : 914087-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172605

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-172605
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart