ENA423210115 Enamine

2-cyano-N-(1-phenylpropyl)acetamide

  • MDL number MFCD07164350
  • Cas Number : 170449-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172634
Availability details for
Quantity
  • Availability
  • Add to Cart