ENA423210130 Enamine

4-bromo-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD24572880
  • Cas Number : 14022-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172656
Availability details for
Quantity
  • Availability
  • Add to Cart