ENA423210138 Enamine

N-benzyl-2-cyano-N-methylacetamide

  • MDL number MFCD09947110
  • Cas Number : 1017040-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172667
Availability details for
Quantity
  • Availability
  • Add to Cart