ENA423210141 Enamine

2-(thiophen-2-yl)ethane-1-sulfonyl chloride

  • MDL number MFCD09886662
  • Cas Number : 405308-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172670
Availability details for
Quantity
  • Availability
  • Add to Cart