ENA423210147 Enamine

tert-butyl N-[2-(aminomethyl)cyclohexyl]carbamate

  • MDL number MFCD19441637
  • Cas Number : 1697833-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171979
Availability details for
Quantity
  • Availability
  • Add to Cart