ENA423210151 Enamine

1,3-diethyl 2-[2-(3-nitrophenyl)hydrazin-1-ylidene]propanedioate

  • MDL number MFCD00623919
  • Cas Number : 13631-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171983

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171983
Availability details for
Quantity
  • Availability
  • Add to Cart