ENA423210154 Enamine

6-bromo-2-methyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD12196496
  • Cas Number : 1159979-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172744
Availability details for
Quantity
  • Availability
  • Add to Cart