ENA423210156 Enamine

[3-(dimethylamino)propyl](propyl)amine

  • MDL number MFCD11146411
  • Cas Number : 19475-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172747
Availability details for
Quantity
  • Availability
  • Add to Cart