ENA423210158 Enamine

4-(2-methanesulfonamidoethyl)benzene-1-sulfonamide

  • MDL number MFCD01097964
  • Cas Number : 349625-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172749

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172749
Availability details for
Quantity
  • Availability
  • Add to Cart