ENA423210169 Enamine

N-benzyl-2-cyano-N-ethylacetamide

  • MDL number MFCD11195405
  • Cas Number : 1039869-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172775
Availability details for
Quantity
  • Availability
  • Add to Cart