ENA423210170 Enamine

1-(4-amino-1,2,5-oxadiazol-3-yl)ethan-1-one

  • MDL number MFCD17015418
  • Cas Number : 17647-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172776
Availability details for
Quantity
  • Availability
  • Add to Cart