ENA423210187 Enamine

4-(3-bromophenoxy)pyrimidine

  • MDL number MFCD14605500
  • Cas Number : 315189-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172911
Availability details for
Quantity
  • Availability
  • Add to Cart