ENA423210192 Enamine

4-(1-methyl-1H-pyrazol-4-yl)-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD17251124
  • Cas Number : 1339381-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172943
Availability details for
Quantity
  • Availability
  • Add to Cart