ENA423210193 Enamine

3-amino-1-[(2,4-difluorophenyl)methyl]piperidin-2-one

  • MDL number MFCD19590674
  • Cas Number : 1404726-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 80
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172944
Availability details for
Quantity
  • Availability
  • Add to Cart