ENA423210194 Enamine

3-(3-methylbutyl)-1H-pyrazol-5-amine

  • MDL number MFCD10692156
  • Cas Number : 1187862-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172945
Availability details for
Quantity
  • Availability
  • Add to Cart