ENA423210211 Enamine

5-(1-chloroethyl)-1-cyclohexyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD24477480
  • Cas Number : 858483-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173019
Availability details for
Quantity
  • Availability
  • Add to Cart