ENA423210220 Enamine

methyl 2-(6-nitro-4-oxo-3,4-dihydroquinazolin-3-yl)acetate

  • MDL number MFCD12635568
  • Cas Number : 67305-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173043
Availability details for
Quantity
  • Availability
  • Add to Cart