ENA423210222 Enamine

[(2,6-difluorophenyl)methyl](methyl)amine

  • MDL number MFCD09736889
  • Cas Number : 392691-62-2
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173048

Safety Information

signalword Danger
pcodes P260,P261,P264,P272,P280,P285,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P333+P313,P342+P311,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-173048
pictograms GHS05,GHS08
hstatements H314,H317,H334
Availability details for
Quantity
  • Availability
  • Add to Cart