ENA423210237 Enamine

3-chloro-N-(1-cyanocyclobutyl)-2,2-dimethylpropanamide

  • MDL number MFCD18856652
  • Cas Number : 1340072-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173110
Availability details for
Quantity
  • Availability
  • Add to Cart