ENA423210242 Enamine

3,4,5,9-tetraazatricyclo[8.4.0.0,3,7]tetradeca-1(10),4,6,11,13-pentaene

  • MDL number MFCD20678312
  • Cas Number : 127933-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173116
Availability details for
Quantity
  • Availability
  • Add to Cart