ENA423210244 Enamine

4-chloro-8-methoxy-3-methylquinoline

  • MDL number MFCD18430858
  • Cas Number : 39593-11-8
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-methyl-8-quinolinyl methyl ether
Inchi Code 1S/C11H10ClNO/c1-7-6-13-11-8(10(7)12)4-3-5-9(11)14-2/h3-6H,1-2H3
Inchi Key UBQXDVNXLJEVKN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173119
Availability details for
Quantity
  • Availability
  • Add to Cart