ENA423210246 Enamine

2-[butyl(methyl)amino]-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-ol

  • MDL number MFCD16216687
  • Cas Number : 1267263-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173124
Availability details for
Quantity
  • Availability
  • Add to Cart