ENA423210250 Enamine

1-(1-benzofuran-2-yl)cyclopropan-1-amine

  • MDL number MFCD17292414
  • Cas Number : 1266172-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171951
Availability details for
Quantity
  • Availability
  • Add to Cart