ENA423210251 Enamine

tert-butyl N-(1-carbamothioylpiperidin-4-yl)carbamate

  • MDL number MFCD24470050
  • Cas Number : 848498-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172041
Availability details for
Quantity
  • Availability
  • Add to Cart