ENA423210252 Enamine

2-methyl-3-(3-nitrophenyl)prop-2-enal

  • MDL number MFCD21851798
  • Cas Number : 37524-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172043
Availability details for
Quantity
  • Availability
  • Add to Cart