ENA423210256 Enamine

6-fluoro-2,3-dimethoxybenzoic acid

  • MDL number MFCD13193706
  • Cas Number : 265670-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173196
Availability details for
Quantity
  • Availability
  • Add to Cart