ENA423210262 Enamine

3-(oxiran-2-yl)benzonitrile

  • MDL number MFCD19597734
  • Cas Number : 13906-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172083
Availability details for
Quantity
  • Availability
  • Add to Cart