ENA423210270 Enamine

[4-(but-2-yn-1-yloxy)phenyl]methanol

  • MDL number MFCD16706009
  • Cas Number : 1250387-44-0
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173240
Availability details for
Quantity
  • Availability
  • Add to Cart