ENA423210277 Enamine

1-(4-nitrophenyl)butane-1,3-dione

  • MDL number MFCD00187226
  • Cas Number : 4023-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173256
Availability details for
Quantity
  • Availability
  • Add to Cart