ENA423210283 Enamine

4-nitro-1H-indol-5-ol

  • MDL number MFCD12924715
  • Cas Number : 135531-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173295
Availability details for
Quantity
  • Availability
  • Add to Cart