ENA423210286 Enamine

3-(4-hydroxyphenyl)-1,2,4-oxadiazol-5-ol

  • MDL number MFCD18872638
  • Cas Number : 1342289-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173303
Availability details for
Quantity
  • Availability
  • Add to Cart