ENA423210288 Enamine

1-(2,6-difluorophenyl)-4-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD27729451
  • Cas Number : 1708740-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172091
Availability details for
Quantity
  • Availability
  • Add to Cart