ENA423210312 Enamine

3-hydroxy-2-(thiophen-2-yl)-7-(trifluoromethyl)-1,4-dihydroquinolin-4-one

  • MDL number MFCD06376666
  • Cas Number : 851268-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-173345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173345
Availability details for
Quantity
  • Availability
  • Add to Cart