ENA423210313 Enamine

1-cyclopropyl-3,3-dimethylbutan-1-one

  • MDL number MFCD20335976
  • Cas Number : 34640-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172092
Availability details for
Quantity
  • Availability
  • Add to Cart