ENA423210319 Enamine

4-methoxypiperidine-1-carbonyl chloride

  • MDL number MFCD23790141
  • Cas Number : 1032707-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173357
Availability details for
Quantity
  • Availability
  • Add to Cart