ENA423210329 Enamine

4-(1H-indol-3-yl)butan-1-amine

  • MDL number MFCD20311166
  • Cas Number : 669-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173433
Availability details for
Quantity
  • Availability
  • Add to Cart