ENA423210339 Enamine

2-amino-N-(2,3-dihydro-1,4-benzodioxin-6-yl)benzene-1-sulfonamide

  • MDL number MFCD13367348
  • Cas Number : 1040334-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172120

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172120
Availability details for
Quantity
  • Availability
  • Add to Cart