ENA423210342 Enamine

3-{6-bromo-3H-imidazo[4,5-b]pyridin-2-yl}pyridine

  • MDL number MFCD00708336
  • Cas Number : 65147-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173552
Availability details for
Quantity
  • Availability
  • Add to Cart