ENA423210343 Enamine

7-methoxypteridin-4-amine

  • MDL number MFCD20705970
  • Cas Number : 89937-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172122
Availability details for
Quantity
  • Availability
  • Add to Cart