ENA423210344 Enamine

2-(2-phenylethyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD14654400
  • Cas Number : 1251377-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173555
Availability details for
Quantity
  • Availability
  • Add to Cart