ENA423210346 Enamine

spiro[3.5]nonan-1-amine hydrochloride

  • MDL number MFCD26407276
  • Cas Number : 1803611-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172123
Availability details for
Quantity
  • Availability
  • Add to Cart